missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD79B Protein

A cDNA sequence encoding the CD79B was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11348-HIS-EC-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15936968

  • € 4090.00 / 1mg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

974
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMARSEDR YRNPKGSACS RIWQSPRFIA RKRGFTVKMH CYMNSASGNV SWLWKQEMDE NPQQLKLEKG RMEESQNESL ATLTIQGIRF EDNGIYFCQQ KCNNTSEVYQ GCGTELRVMG FSTLAQLKQR NTLKD
Greater than 90% as determined by SDS-PAGE.
CD79B Protein
Research Use Only
CD79B
Human
His
CD79B protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 2M UREA and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers