missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD8A / MAL Protein
A DNA sequence encoding the Homo sapiens (Human) CD8A / MAL, was expressed in the hosts and tags indicated.
€ 202.00 - € 2155.00
Specifications
Accession Number | NP_001139345.1 |
---|---|
Gene ID (Entrez) | 925 |
Name | CD8A / MAL Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15935549
|
enQuireBio™
QP2208-YE-100UG |
100 μg |
€ 645.00
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975549
|
enQuireBio™
QP2208-YE-500UG |
500 μg |
€ 1740.00
500µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945549
|
enQuireBio™
QP2208-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15965549
|
enQuireBio™
QP2208-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15985549
|
enQuireBio™
QP2208-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15955549
|
enQuireBio™
QP2208-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001139345.1 | |
CD8A / MAL Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
925 | |
Research Use Only | |
CD8A | |
Recombinant Protein | |
Yeast | |
SQFRVSPLDR TWNLGETVEL KCQVLLSNPT SGCSWLFQPR GAAASPTFLL YLSQNKPKAA EGLDTQRFSG KRLGDTFVLT LSDFRRENEG YYFCSALSNS IMYFSHFVPV FLPAKPTTTP APRPPTPAPT IASQPLSLRP EACRPAAGGA VHTRGLDFAC D | |
Greater than 90% as determined by SDS-PAGE. |