missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CDK4 Protein

A DNA sequence encoding the Homo sapiens (Human) CDK4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP10035-ba-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15942788

  • € 1540.00 / 100µg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CDK4 Protein
Research Use Only
CDK4
Recombinant Protein
Baculovirus
MATSRYEPVA EIGVGAYGTV YKARDPHSGH FVALKSVRVP NGGGGGGGLP ISTVREVALL RRLEAFEHPN VVRLMDVCAT SRTDREIKRC FVESLSSVET LKPTSWAKSL T
Greater than 90% as determined by SDS-PAGE.
100 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers