missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CEACAM-1/CD66a Fc Chimera Avi Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human CEACAM-1/CD66a (Catalog # 2244-CM ) is immobilized at 0.5 μg/mL (100 μL/well), Biotinylated Recombinant Human CEACAM-1/CD66a Fc Chimera Avi-tag (Catalog # AVI11074) binds with an ED50 of 5.00-50.0 ng/mL.
Specifications
Specifications
| Accession Number | P13688.2 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 634 |
| Molecular Weight (g/mol) | M.W.-observed: 115-135 kDa, under reducing conditions., M.W.-theroretical: 72 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human CEACAM-1/CD66a protein Human CEACAM-1 (Gln35-Gly428) (C-terminus) Accession # P13688.2 IEGRMD Human IgG1 (Pro100-Lys330) Avi-tag (N-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.50 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | antigen CD66, BGP1, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, Biliary Glycoprotein 1, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen, Cea-1, CEACAM1, Hv-1, Hv-2, MHVR |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction