missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cell death activator CIDE-3 Protein

Product Code. 16071541
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16071541 100 μg 100µg
16081541 10 μg 10µg
16021551 50 μg 50µg
16001551 200 μg 200µg
16011551 500 μg 500µg
16091541 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16071541 Supplier enQuireBio™ Supplier No. QP7912ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Cell death activator CIDE-3, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001186480.1
Gene ID (Entrez) 63924
Name Cell death activator CIDE-3 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol CIDEC
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag GST
Sequence MEYAMKSLSL LYPKSLSRHV SVRTSVVTQQ LLSEPSPKAP RARPCRVSTA DRSVRKGIMA YSLEDLLLKV RDTLMLADKP FFLVLEEDGT TVETEEYFQA LAGDTVFMVL QKGQKWQPPS EQGTRHPLSL SHKPAKKIDV ARVTFDLYKL NPQDFIGCLN VKATFYDTYS LSYDLHCCGA KRIMKEAFRW ALFSMQATGH VLLGTSCYLQ QLLDATEEGQ PPKGKASSLI PTCLKILQ
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.