missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CENPM Protein
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
Specifications
Specifications
| Name | CENPM Protein |
| Quantity | 5 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSMSVLRPL DKLPGLNTAT ILLVGTEDAL LQQLADSMLK EDCASELKVH LAKSLPLPSS VNRPRIDLIV FVVNLHSKYS LQNTEESLRH VDASFFLGKV CFLATGAGRE SHCSIHRHTV VKLAHTYQSP LLYCDLEVEG FRATMAQRLV RVLQICAGHV PGVSALNLLS LLRSSEGPSL EDL |
| Buffer | The CENPM solution contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 30% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction