missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Charged multivesicular body protein 1b Protein

A DNA sequence encoding the Homo sapiens (Human) Charged multivesicular body protein 1b, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7751-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16030581

  • € 2385.00 / 1mg
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_065145.2
Charged multivesicular body protein 1b Protein
Research Use Only
CHMP1B
Recombinant Protein
E. coli
MSNMEKHLFN LKFAAKELSR SAKKCDKEEK AEKAKIKKAI QKGNMEVARI HAENAIRQKN QAVNFLRMSA RVDAVAARVQ TAVTMGKVTK SMAGVVKSMD ATLKTMNLEK ISALMDKFEH QFETLDVQTQ QMEDTMSSTT TLTTPQNQVD MLLQEMADEA GLDLNMELPQ GQTGSVGTSV ASAEQDELSQ RLARLRDQV
Greater than 80% as determined by SDS-PAGE.
57132
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers