missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Checkpoint protein HUS1 Protein
A DNA sequence encoding the Homo sapiens (Human) Checkpoint protein HUS1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6186-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004498.1 | |
Checkpoint protein HUS1 Protein | |
Research Use Only | |
HUS1 | |
Recombinant Protein | |
E. coli | |
KFRAKIVDGA CLNHFTRISN MIAKLAKTCT LRISPDKLNF ILCDKLANGG VSMWCELEQE NFFNEFQMEG VSAENNEIYL ELTSENLSRA LKTAQNARAL KIKLTNKHFP CLTVSVELLS MSSSSRIVTH DIPIKVIPRK LWKDLQEPVV PDPDVSIYLP VLKTMKSVVE KMKNISNHLV IEANLDGELN LKIETELVCV TTHFKDLGNP PLASESTHED RNVEHMAEVH IDIRKLLQFL AGQQVNPTKA LCNIVNNKMV HFDLLHEDVS LQYFIPALS | |
Greater than 90% as determined by SDS-PAGE. |
3364 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |