missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CHI3L2 / YKL-39 Protein
A DNA sequence encoding the Homo sapiens (Human) CHI3L2 / YKL-39, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7515-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001020368.1 | |
CHI3L2 / YKL-39 Protein | |
Research Use Only | |
CHI3L2 | |
Recombinant Protein | |
E. coli | |
YKLVCYFTNW SQDRQEPGKF TPENIDPFLC SHLIYSFASI ENNKVIIKDK SEVMLYQTIN SLKTKNPKLK ILLSIGGYLF GSKGFHPMVD SSTSRLEFIN SIILFLRNHN FDGLDVSWIY PDQKENTHFT VLIHELAEAF QKDFTKSTKE RLLLTAGVSA GRQMIDNSYQ VEKLAKDLDF INLLSFDFHG SWEKPLITGH NSPLSKGWQD RGPSSYYNVE YAVGYWIHKG MPSEKVVMGI PTYGHSFTLA SAETTVGAPA SGPGAAGPIT ESSGFLAYYE ICQFLKGAKI TRLQDQQVPY AVKGNQWVGY DDVKSMETKV QFLKNLNLGG AMIWSIDMDD FTGKSCNQGP YPLVQAVKRS LGSL | |
Greater than 90% as determined by SDS-PAGE. |
1117 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |