missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Chitinase 3-like 1/YKL-40 Protein

Product Code. 18390258
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18390258

Brand: R&D Systems™ 2599CH050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by the ability of the immobilized protein to support the adhesion of FaDu human squamous cell carcinoma cells. When 5 x 104 cells per well are added to recombinant human CHI3-L1 coated plate, cell adhesion is enhanced in a dose dependent manner. The ED50 for this effect is 0.6-2.4 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P36222
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 1116
Molecular Weight (g/mol) M.W. (Observed): 38-41 kDa, reducing conditions; M.W. (theoretical): 41.3 kDa (monomer)
Name Chitinase 3-like 1
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human Chitinase 3-like 1 protein Tyr22-Thr383, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol CHI3L1
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 200 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein Chitinase 3-like 1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.