missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Chitinase domain-containing protein 1

Product Code. p-7154865
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16063181 10 μg 10µg
16003191 50 μg 50µg
16053181 100 μg 100µg
16083181 200 μg 200µg
16093181 500 μg 500µg
16073181 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16063181 Supplier enQuireBio™ Supplier No. QP8193ec10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Chitinase domain-containing protein 1, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001136146.1
Gene ID (Entrez) 66005
Name Chitinase domain-containing protein 1
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Chid1
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence TLSKSDAKKA ASKTLLEKSQ FSDKPVQDRG LVVTDLKAES VVLEHRSYCS AKARDRHFAG DVLGYVTPWN SHGYDVTKVF GSKFTQISPV WLQLKRRGRE MFEVTGLHDV DQGWMRAVRK HAKGLHIVPR LLFEDWTYDD FRNVLDSEDE IEELSKTVVQ VAKNQHFDGF VVEVWNQLLS QKRVGLIHML THLAEALHQA RLLALLVIPP AITPGTDQLG MFTHKEFEQL APVLDGFSLM TYDYSTAHQP GPNAPLSWVR ACVQVLDPKS KWRSKILLGL NFYGMDYATS KDAREPVVGA RYIQTLKDHR PRMVWDSQAS EHFFEYKKSR SGRHVVFYPT LKSLQVRLEL ARELGVGVSI WELGQGLDYF YDLL
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.