missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Chordin-Like 2 Protein

Product Code. 18735557
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18735557

Brand: R&D Systems™ 9127CH050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Chordin-Like 2 Protein is derived from NS0. The Recombinant Human Chordin-Like 2 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6WN34
For Use With (Application) Bioactivity
Gene ID (Entrez) 25884
Molecular Weight (g/mol) Observed M.W.: 44-65kDa, reducing conditions, Theoretical M.W.: 46kDa
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived human Chordin-like 2/CHRDL2 protein Leu21-Thr429,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias BNF-1, Chordin like 2, chordin-like 2, chordin-like protein 2, CHRDL2
Common Name Chordin-like 2/CHRDL2
Gene Symbol CHRDL2
Biological Activity Measured by its ability to inhibit BMP-4-induced activity in MC3T3-E1 mouse preosteoblast cells. The ED50 for this effect is typically 0.4-2.4μg/mL
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Leu21-Thr429, with a C-terminal 6-His tag
Purity or Quality Grade >85%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.