missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CLEC10A Protein
A cDNA sequence encoding the CLEC10A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11442-1mg
Additional Details : Weight : 0.01000kg
Specifications
CLEC10A Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADPQNSKFQR DLVTLRTDFS NFTSNTVAEI QALTSQGSSL EETIASLKAE VEGFKQERQA VHSEMLLRVQ QLVQDLKKLT CQVATLNNNG EEASTEGTCC PVNWVEHQDS CYWFSHSGMS WAEAEKYCQL KNAHLVVINS REEQNFVQKY LGSAYTWMGL SDPEGAWKWV DGTDYATGFQ NWKPGQPDDW QGHGLGGGED CAHFHPDGRW NDDVCQRPYH WVCEAGLGQT SQESHHHHHH H | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CLEC10A protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4). |