missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CLEC1B Protein

Product Code. 15967278
Change view
Click to view available options
Quantity:
1 μg
1 mg
20 μg
5 μg
50 μg
Unit Size:
1mg
1µg
20µg
50µg
5µg
Product Code. Quantity unitSize
15967278 1 μg 1µg
15997278 5 μg 5µg
15977278 20 μg 20µg
15987278 50 μg 50µg
15957278 1 mg 1mg
5 options
This item is not returnable. View return policy

Product Code. 15967278

Brand: enQuireBio™ QP11443HIS1ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the CLEC1B was constructed and used to recombinantly synthesize the protein.

Specifications

Name CLEC1B Protein
Quantity 1 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGSSVMQRNY LQGENENRTG TLQQLAKRFC QYVVKQSELK GTFKGHKCSP CDTNWRYYGD SCYGFFRHNL TWEESKQYCT DMNATLLKID NRNIVEYIKA RTHLIRWVGL SRQKSNEVWK WEDGSVISEN MFEFLEDGKG NMNCAYFHNG KMHPTFCENK HYLMCERKAG MTKVDQLP
Buffer CLEC1B protein solution (1 mg/ml) containing 20mM Tris, PH 8.0 and 10% glycerol.
Purity or Quality Grade Greater than 90.0% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.