missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CNTF Protein
Click to view available options
Quantity:
1 mg
500 μg
Unit Size:
1mg
500µg
Specifications
Specifications
| Accession Number | NP_000605.1 |
| Gene ID (Entrez) | 1270 |
| Name | CNTF Protein |
| Quantity | 500 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
| Gene Symbol | CNTF |
| Biological Activity | Shows bioactivity at a concentration of 325 ng/mL in the TF-1 cell proliferation assay. The bioactivity acceptance criteria is set at 3.1 x 10^3 units/mg. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction