missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Coactosin-like Protein
A DNA sequence encoding the Homo sapiens (Human) Coactosin-like protein, was expressed in the hosts and tags indicated.
€ 160.00 - € 2385.00
Specifications
Accession Number | NP_066972.1 |
---|---|
Gene ID (Entrez) | 23406 |
Name | Coactosin-like Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16089620
|
enQuireBio™
QP7594-EC-100UG |
100 μg |
€ 530.00
100µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16009630
|
enQuireBio™
QP7594-EC-1MG |
1 mg |
€ 2385.00
1mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16099620
|
enQuireBio™
QP7594-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16019630
|
enQuireBio™
QP7594-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16029630
|
enQuireBio™
QP7594-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16039630
|
enQuireBio™
QP7594-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_066972.1 | |
Coactosin-like Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
23406 | |
Research Use Only | |
COTL1 | |
Recombinant Protein | |
E. coli | |
ATKIDKEACR AAYNLVRDDG SAVIWVTFKY DGSTIVPGEQ GAEYQHFIQQ CTDDVRLFAF VRFTTGDAMS KRSKFALITW IGENVSGLQR AKTGTDKTLV KEVVQNFAKE FVISDRKELE EDFIKSELKK AGGANYDAQT E | |
Greater than 90% as determined by SDS-PAGE. |