missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Coagulation factor V Protein
A DNA sequence encoding the Homo sapiens (Human) Coagulation factor V, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6011-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000121.2 | |
Coagulation factor V Protein | |
Research Use Only | |
F5 | |
Recombinant Protein | |
E. coli | |
MPSPSSPTLN DTFLSKEFNP LVIVGLSKDG TDYIEIIPKE EVQSSEDDYA EIDYVPYDDP YKTDVRTNIN SSRDPDNIAA WYLRSNNGNR RNYYIAAEEI SWDYSEFVQR ETDIEDSDDI PEDTT | |
Greater than 90% as determined by SDS-PAGE. |
2153 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |