missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Complement Component C5a Protein
Alle ansehen Bio Techne Produkte
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Packungsgröße:
100µg
1mg
500µg
5µg
Beschreibung
Fully biologically active when compared to standard. The biological activity determined by its ability to chemoattract human monocytes using a concentration range of 1.0-10.0 ng/mL.
Spezifikation
Spezifikation
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 727 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 8.3 kDa |
| Quantity | 5 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin Concentration | Less than 1 EU/μg of Complement Component C5a as determined by LAL method. |
| Gene Alias | C5, C5A, HE |
| Gene Symbol | C5 |
| Reconstitution | Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.) |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur