Learn More
enQuireBio™ Recombinant Human Complexin-2 / CPLX2 Protein
Product Code. 16090701
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Product Code. 16090701
Brand: enQuireBio™ QP7773ec1mg
A DNA sequence encoding the Homo sapiens (Human) Complexin-2 / CPLX2, was expressed in the hosts and tags indicated.
Specifications
Accession Number NP_001008221.1
Gene ID (Entrez) 10814
Name Complexin-2 / CPLX2 Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol CPLX2
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag GST
Sequence MDFVMKQALG GATKDMGKML GGEEEKDPDA QKKEEERQEA LRQQEEERKA KHARMEAERE KVRQQIRDKY GLKKKEEKEA EEKAALEQPC EGSLTRPKKA IPAGCGDEEE EEEESILDTV LKYLPGPLQD MFKK
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.