missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Craniofacial development protein 1
A DNA sequence encoding the Homo sapiens (Human) Craniofacial development protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5824-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006315.1 | |
Craniofacial development protein 1 | |
Research Use Only | |
CFDP1 | |
Recombinant Protein | |
E. coli | |
MEEFDSEDFS TSEEDEDYVP SGGEYSEDDV NELVKEDEVD GEEQTQKTQG KKRKAQSIPA RKRRQGGLSL EEEEEEDANS ESEGSSSEEE DDAAEQEKGI GSEDARKKKE DELWASFLND VGPKSKVPPS TQVKKGEETE ETSSSKLLVK AEELEKPKET EKVKITKVFD FAGEEVRVTK EVDATSKEAK SFFKQNEKEK PQANVPSALP SLPAGSGLKR SSGMSSLLGK IGAKKQKMST LEKSKLDWES FKEEEGIGEE LAIHNRGKEG YIERKAFLDR VDHRQFEIER DLRLSKMKP | |
Greater than 80% as determined by SDS-PAGE. |
10428 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |