missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CRYAA Protein
Klik for at se tilgængelige muligheder
Quantity:
1 mg
100 μg
20 μg
Pakningsstørrelse:
100µg
1mg
20µg
Tekniske data
Tekniske data
| Name | CRYAA Protein |
| Quantity | 20 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | Untagged |
| Sequence | MDVTIQHPWF KRTLGPFYPS RLFDQFFGEG LFEYDLLPFL SSTISPYYRQ SLFRTVLDSGISEVRSDRDK FVIFLDVKHF SPEDLTVKVQ DDFVEIHGKH NERQDDHGYI SREFHRRYRLPSNVDQSALS CSLSADGMLT FCGPKIQTGL DATHAERAIP VSREEKPTSA PSS |
| Buffer | The CRYAA protein contains 20mM Tris-HCl buffer (pH 7.5), 50mM NaCl and 1mM EDTA. |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion