missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CRYAA Protein

Product Code. 15917508
Click to view available options
Quantity:
1 mg
100 μg
20 μg
Unit Size:
100µg
1mg
20µg
This item is not returnable. View return policy

Product Code. 15917508

Brand: enQuireBio™ QP115171mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the CRYAA was constructed and used to recombinantly synthesize the protein.

Specifications

Name CRYAA Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence MDVTIQHPWF KRTLGPFYPS RLFDQFFGEG LFEYDLLPFL SSTISPYYRQ SLFRTVLDSGISEVRSDRDK FVIFLDVKHF SPEDLTVKVQ DDFVEIHGKH NERQDDHGYI SREFHRRYRLPSNVDQSALS CSLSADGMLT FCGPKIQTGL DATHAERAIP VSREEKPTSA PSS
Buffer The CRYAA protein contains 20mM Tris-HCl buffer (pH 7.5), 50mM NaCl and 1mM EDTA.
Purity or Quality Grade Greater than 95.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.