missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human CTLA-4 Fc Chimera (CHO-expressed)

Product Code. 18137630
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 18137630

Brand: R&D Systems™ 7268CT100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to inhibit IL-2 secretion by stimulated Jurkat human acute T cell leukemia cells. Linsley, P.S. et al. (1991) J. Exp. Med. 174:561. The ED50 for this effect is 0.03-0.15 μg/mL when stimulated with 1 μg/mL Recombinant Human B7-1/CD80 Fc Chimera (Catalog # 140-B1 ).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P16410
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 1493
Molecular Weight (g/mol) M.W. (Observed): 53-58 kDa, reducing conditions; M.W. (theoretical): 40.1 kDa (monomer)
Name CTLA-4 Fc Chimera
Quantity 100 μg
Source Chinese Hamster Ovary cell line, CHO-derived human CTLA-4 protein Human CTLA-4 (Ala37-Phe162) (N-terminus) Accession # P16410 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol CTLA4
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein CTLA-4
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.