Learn More
DiscoverX Recombinant Human CXCL11
Product Code. 17322359
Encompass
Click to view available options
:
5 μg
Product Code. 17322359
Brand: DiscoverX 921010
Recombinant Human CXCL11 is an agonist for CXCR3. Provided as 5μg of dry powder alongside reconstitution buffer.
CXCL11 is a small cytokine protein that acts as a chemoattractant for IL-2-activated T cells, but does not affect freshly isolated unstimulated T cells, neutrophils, or monocytes. CXCL11 is a non-ELR chemokine that is regulated by interferons and elicits its effects by acting as an agonist for the CXCR3 receptor. Provided as 5μg of dry powder alongside reconstitution buffer.
Specifications
Quantity 5 μg
Product Type Control Ligands