Learn More
DiscoverX Recombinant Human CXCL11
Click to view available options
:
5 μg
Description
CXCL11 is a small cytokine protein that acts as a chemoattractant for IL-2-activated T cells, but does not affect freshly isolated unstimulated T cells, neutrophils, or monocytes. CXCL11 is a non-ELR chemokine that is regulated by interferons and elicits its effects by acting as an agonist for the CXCR3 receptor. Provided as 5μg of dry powder alongside reconstitution buffer.
Specifications
Specifications
| Quantity | 5 μg |
| Product Type | Control Ligands |