Learn More
DiscoverX Recombinant Human CXCL16
Product Code. 17342359
Encompass
Click to view available options
:
5 μg
Product Code. 17342359
Brand: DiscoverX 921013
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Recombinant Human CXCL16 is an agonist for CXCR6. Provided as 5μg of dry powder alongside reconstitution buffer.
CXCL16 is a cytokine protein expressed as a cell surface bound molecule, as well as a soluble chemokine, by macrophages, monocytes, B cells, and Schwann cells. Its expression is induced by IFNG and TNF-α. The soluble protein is chemotactic for activated CD8 T cells, inducing the process of calcium flux and regulating their migration. CXCL16 is an agonist for the CXCR6 receptor, but also serves as a receptor for oxidized LDL and phosphadtidylserine. Provided as 5μg of dry powder alongside reconstitution buffer.
Specifications
Quantity 5 μg
Product Type Control Ligands