missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cytokeratin 8 Protein

Product Code. 16032180
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Product Code. Quantity unitSize
16032180 1 mg 1mg
16022180 10 μg 10µg
16062180 50 μg 50µg
16012180 100 μg 100µg
16042180 200 μg 200µg
16052180 500 μg 500µg
6 missing translation for 'options'
This item is not returnable. View return policy

Product Code. 16032180

Brand: enQuireBio™ QP6284ec1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Cytokeratin 8, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001243222.1
Gene ID (Entrez) 3856
Name Cytokeratin 8 Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol KRT8
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence SIRVTQKSYK VSTSGPRAFS SRSYTSGPGS RISSSSFSRV GSSNFRGGLG GGYGGASGMG GITAVTVNQS LLSPLVLEVD PNIQAVRTQE KEQIKTLNNK FASFIDKVRF LEQQNKMLET KWSLLQQQKT ARSNMDNMFE SYINNLRRQL ETLGQEKLKL EAELGNMQGL VEDFKNKYED EINKRTEMEN EFVLIKKDVD EAYMNKVELE SRLEGLTDEI NFLRQLYEEE IRELQSQISD TSVVLSMDNS RSLDMDSIIA EVKAQYEDIA NRSRAEAESM YQIKYEELQS LAGKHGDDLR RTKTEISEMN RNISRLQAEI EGLKGQRASL EAAIADAEQR GELAIKDANA KLSELEAALQ RAKQDMARQL REYQELMNVK LALDIEIATY RKLLEGEESR LESGMQNMSI HTKTTSGYAG GLSSAYGGLT SPGLSYSLGS SFGSGAGSSS FSRTSSSRAV VVKKIETRDG KLVSESSDVL PK
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.