missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human D-amino acid oxidase activator Protein
A DNA sequence encoding the Homo sapiens (Human) D-amino acid oxidase activator, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5911-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001155284.1 | |
D-amino acid oxidase activator Protein | |
Research Use Only | |
DAOA | |
Recombinant Protein | |
E. coli | |
MLEKLMGADS LQLFRSRYTL GKIYFIGFQR SILLSKSENS LNSIAKETEE GRETVTRKEG WKRRHEDGYL EMAQRHLQRS LCPWVSYLPQ PYAELEEVSS HVGKVFMARN YEFLAYEASK DRRQPLERMW TCNYNQQKDQ SCNHKEITST KAE | |
Greater than 90% as determined by SDS-PAGE. |
267012 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |