missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DBI / ACBD1 Protein

A DNA sequence encoding the Homo sapiens (Human) DBI / ACBD1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP1027-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15933308

  • € 212.00 / 10µg
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001073331.1
DBI / ACBD1 Protein
Research Use Only
DBI
Recombinant Protein
E. coli
SQAEFEKAAE EVRHLKTKPS DEEMLFIYGH YKQATVGDIN TERPGMLDFT GKAKWDAWNE LKGTSKEDAM KAYINKVEEL KKKYGI
Greater than 90% as determined by SDS-PAGE.
1622
10 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers