missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DCXR / HCR2 Protein

A DNA sequence encoding the Homo sapiens (Human) DCXR / HCR2, was expressed in the hosts and tags indicated.

€ 160.00 - € 2385.00

Specifications

Accession Number NP_001182147.1
Gene ID (Entrez) 51181
Name DCXR / HCR2 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16030831
View Documents Promotion Details
enQuireBio™
QP7792-EC-100UG
100 μg
€ 530.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16050831
View Documents Promotion Details
enQuireBio™
QP7792-EC-1MG
1 mg
€ 2385.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16040831
View Documents Promotion Details
enQuireBio™
QP7792-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16060831
View Documents Promotion Details
enQuireBio™
QP7792-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16070831
View Documents Promotion Details
enQuireBio™
QP7792-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16080831
View Documents Promotion Details
enQuireBio™
QP7792-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_001182147.1
DCXR / HCR2 Protein
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
51181
Research Use Only
DCXR
Recombinant Protein
E. coli
MELFLAGRRV LVTGAGKGIG RGTVQALHAT GARVVAVSRT QADLDSLVRE CPGIEPVCVD LGDWEATERA LGSVGPVDLL VNNAAVALLQ PFLEVTKEAF DRSFEVNLRA VIQVSQIVAR GLIARGVPGA IVNVSSQCSQ RAVTNHSVYC STKGALDMLT KVMALELGPH KIRVNAVNPT VVMTSMGQAT WSDPHKAKTM LNRIPLGKFA EVEHVVNAIL FLLSDRSGMT TGSTLPVEGG FWAC
Greater than 80% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications