missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DCXR / HCR2 Protein

A DNA sequence encoding the Homo sapiens (Human) DCXR / HCR2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7792-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16030831

  • € 530.00 / 100µg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001182147.1
DCXR / HCR2 Protein
Research Use Only
DCXR
Recombinant Protein
E. coli
MELFLAGRRV LVTGAGKGIG RGTVQALHAT GARVVAVSRT QADLDSLVRE CPGIEPVCVD LGDWEATERA LGSVGPVDLL VNNAAVALLQ PFLEVTKEAF DRSFEVNLRA VIQVSQIVAR GLIARGVPGA IVNVSSQCSQ RAVTNHSVYC STKGALDMLT KVMALELGPH KIRVNAVNPT VVMTSMGQAT WSDPHKAKTM LNRIPLGKFA EVEHVVNAIL FLLSDRSGMT TGSTLPVEGG FWAC
Greater than 80% as determined by SDS-PAGE.
51181
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers