missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Death effector domain-containing Protein

A DNA sequence encoding the Homo sapiens (Human) Death effector domain-containing protein, was expressed in the hosts and tags indicated.

€ 160.00 - € 2385.00

Specifications

Accession Number NP_001034800.1
Gene ID (Entrez) 9191
Name Death effector domain-containing Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15918709
View Documents Promotion Details
enQuireBio™
QP5923-EC-100UG
100 μg
€ 530.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15938709
View Documents Promotion Details
enQuireBio™
QP5923-EC-1MG
1 mg
€ 2385.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15928709
View Documents Promotion Details
enQuireBio™
QP5923-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15948709
View Documents Promotion Details
enQuireBio™
QP5923-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15958709
View Documents Promotion Details
enQuireBio™
QP5923-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15968709
View Documents Promotion Details
enQuireBio™
QP5923-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_001034800.1
Death effector domain-containing Protein
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
9191
Research Use Only
DEDD
Recombinant Protein
E. coli
MAGLKRRASQ VWPEEHGEQE HGLYSLHRMF DIVGTHLTHR DVRVLSFLFV DVIDDHERGL IRNGRDFLLA LERQGRCDES NFRQVLQLLR IITRHDLLPY VTLKRRRAVC PDLVDKYLEE TSIRYVTPRA LSDPEPRPPQ PSKTVPPHYP VVCCPTSGPQ MCSKRPARGR ATLGSQRKRR KSVTPDPKEK QTCDIRLRVR AEYCQHETAL QGNVFSNKQD PLERQFERFN QANTILKSRD LGSIICDIKF SELTYLDAFW RDYINGSLLE ALKGVFITDS LKQAVGHEAI KLLVNVDEED YELGRQKLLR NLMLQALP
Greater than 80% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications