missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DEFB116 Protein
A cDNA sequence encoding the DEFB116 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11617-1mg
Additional Details : Weight : 0.01000kg
Specifications
245930 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSGLFRSHN GKSREPWNPC ELYQGMCRNA CREYEIQYLT CPNDQKCCLK LSVKITSSKN VKEDYDSNSN LSVTNSSSYS HI | |
Greater than 80.0% as determined by SDS-PAGE. |
DEFB116 Protein | |
Research Use Only | |
DEFB116 | |
Human | |
His | |
DEFB116 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |