missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Deoxyguanosine kinase, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) Deoxyguanosine kinase, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7588-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16039590

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001305788.1
Deoxyguanosine kinase, mitochondrial Protein
Research Use Only
DGUOK
Recombinant Protein
E. coli
RLSIEGNIAV GKSTFVKLLT KTYPEWHVAT EPVATWQNIQ AAGTQKACTA QSLGNLLDMM YREPARWSYT FQTFSFLSRL KVQLEPFPEK LLQARKPVQI FERSVYSDRY IFAKNLFENG SLSDIEWHIY QDWHSFLLWE FASRITLHGF IYLQASPQVC LKRLYQRARE EEKGIELAYL EQLHGQHEAW LIHKTTKLHF EALMNIPVLV LDVNDDFSEE VTKQEDLMRE VNTFVKNL
Greater than 90% as determined by SDS-PAGE.
1716
10 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers