missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Deoxyguanosine kinase, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) Deoxyguanosine kinase, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7588-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16029590

  • € 406.00 / 100µg


Estimated Shipment
23-10-2020
Add to basket

Specifications

Specifications

NP_001305788.1
Deoxyguanosine kinase, mitochondrial Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
1716
Greater than 90% as determined by SDS-PAGE.
Research Use Only
DGUOK
Recombinant Protein
E. coli
RLSIEGNIAV GKSTFVKLLT KTYPEWHVAT EPVATWQNIQ AAGTQKACTA QSLGNLLDMM YREPARWSYT FQTFSFLSRL KVQLEPFPEK LLQARKPVQI FERSVYSDRY IFAKNLFENG SLSDIEWHIY QDWHSFLLWE FASRITLHGF IYLQASPQVC LKRLYQRARE EEKGIELAYL EQLHGQHEAW LIHKTTKLHF EALMNIPVLV LDVNDDFSEE VTKQEDLMRE VNTFVKNL
Videos
Documents