missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DHRS2 Protein
A DNA sequence encoding the Homo sapiens (Human) DHRS2, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP7548-ec-10ug
Additional Details : Weight : 0.01000kg
Spezifikation
NP_005785.1 | |
DHRS2 Protein | |
Research Use Only | |
DHRS2 | |
Recombinant Protein | |
E. coli | |
MLSAVARGYQ GWFHPCARLS VRMSSTGIDR KGVLANRVAV VTGSTSGIGF AIARRLARDG AHVVISSRKQ QNVDRAMAKL QGEGLSVAGI VCHVGKAEDR EQLVAKALEH CGGVDFLVCS AGVNPLVGST LGTSEQIWDK ILSVNVKSPA LLLSQLLPYM ENRRGAVILV SSIAAYNPVV ALGVYNVSKT ALLGLTRTLA LELAPKDIRV NCVVPGIIKT DFSKVVRIGF MGMSLSGRTS RNIISCRGLG SQRTVQESCP SCALQMPATS TGRTLRWQAT PLGSERSGGG CVAVVPGPGA | |
Greater than 80% as determined by SDS-PAGE. |
10202 | |
10 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |