missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Dihydrolipoamide Dehydrogenase/DLD

Product Code. 18177660
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18177660 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18177660 Supplier R&D Systems™ Supplier No. 8646DH050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Dihydrolipoamide Dehydrogenase/DLD is derived from E. coli. The Recombinant Human Dihydrolipoamide Dehydrogenase/DLD has been validated for the following applications: Enzyme Activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P09622
For Use With (Application) Enzyme Activity
Gene ID (Entrez) 1738
Molecular Weight (g/mol) Observed M.W.: 52-62kDa, reducing conditions, Theoretical M.W.: 52kDa
Quantity 50 μg
Source E. coli-derived human Dihydrolipoamide Dehydrogenase/DLD protein GGS HHHHHH GMASLENLYFQ Human DLD (Ala36-Phe509) Accession &Num; P09622 N-terminus C-terminusAla36-Phe509,with N-terminal GGS-6-His tag and GMASLENLYFQ insertion
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method.
Gene Alias 2-oxo-glutarate complex, branched chain keto acid dehydrogenase complex), branched chain keto acid dehydrogenase complex, Diaphorase, dihydrolipoamide dehydrogenase (E3 component of pyruvate dehydrogenase complex, dihydrolipoamide dehydrogenasemitochondrial, DLD, DLDD, DLDH, E3, E3 component of pyruvate dehydrogenase complex, 2-oxo-glutarate complex, EC 1.8.1, EC 1.8.1.4, GCSL, Glycine cleavage system L protein, glycine cleavage system protein L, LAD, Lipoamide Dehydrogenase, lipoamide reductase, lipoyl dehydrogenase, PHE3
Common Name Dihydrolipoamide Dehydrogenase/DLD
Gene Symbol DLD
Biological Activity Measured by its ability to produce NADH during the oxidation of lipoic acid. The specific activity is >5,000 pmol/min/μg, as measured under the described conditions. See Activity Assay Protocol on www.RnDSystems.com.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System E. coli-derived GGS, N-terminus; HHHHHH; GMASLENLYFQ; Human DLD (Ala36-Phe509) Accession # P09622, C-terminus (Ala36 - Phe509, with N-terminal GGS-6-His tag and GMASLENLYFQ insertion)
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5μg per lane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.