missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DNA repair protein XRCC4 Protein

A DNA sequence encoding the Homo sapiens (Human) DNA repair protein XRCC4, was expressed in the hosts and tags indicated.

€ 245.00 - € 3405.00

Specifications

Accession Number NP_001304941.1
Gene ID (Entrez) 7518
Name DNA repair protein XRCC4 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16089260
View Documents Promotion Details
enQuireBio™
QP7540-EC-100UG
100 μg
€ 770.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16009270
View Documents Promotion Details
enQuireBio™
QP7540-EC-1MG
1 mg
€ 3405.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16099260
View Documents Promotion Details
enQuireBio™
QP7540-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16019270
View Documents Promotion Details
enQuireBio™
QP7540-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16029270
View Documents Promotion Details
enQuireBio™
QP7540-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16039270
View Documents Promotion Details
enQuireBio™
QP7540-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_001304941.1
DNA repair protein XRCC4 Protein
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
7518
Research Use Only
XRCC4
Recombinant Protein
E. coli
MERKISRIHL VSEPSITHFL QVSWEKTLES GFVITLTDGH SAWTGTVSES EISQEADDMA MEKGKYVGEL RKALLSGAGP ADVYTFNFSK ESCYFFFEKN LKDVSFRLGS FNLEKVENPA EVIRELICYC LDTIAENQAK NEHLQKENER LLRDWNDVQG RFEKCVSAKE ALETDLYKRF ILVLNEKKTK IRSLHNKLLN AAQEREKDIK QEGETAICSE MTADRDPVYD ESTDEESENQ TDLSGLASAA VSKDDSIISS LDVTDIAPSR KRRQRMQRNL GTEPKMAPQE NQLQEKENSR PDSSLPETSK KEHISAENMS LETLRNSSPE DLFDEI
Greater than 80% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications