missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DNAJC27 Protein

A cDNA sequence encoding the DNAJC27 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11681-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15928028

  • € 7875.00 / 1mg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

DNAJC27 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMEANMPK RKEPGRSLRI KVISMGNAEV GKSCIIKRYC EKRFVSKYLA TIGIDYGVTK VHVRDREIKV NIFDMAGHPF FYEVRNEFYK DTQGVILVYD VGQKDSFDAL DAWLAEMKQE LGPHGNMENI IFVVCANKID CTKHRCVDES EGRLWAESKG FLYFETSAQT GEGINEMFQT FYISIVDLCE NGGKRPTTNS SASFTKEQAD AIRRIRNSKD SWDMLGVKPG ASRDEVNKAY RKLAVLLHPD KCVAPGSEDA FKAVVNARTA LLKNIK
Greater than 90% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The DNAJC27 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol, 2mM DTT and 2mM EDTA.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers