missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNAJC27 Protein
A cDNA sequence encoding the DNAJC27 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11681-2ug
Additional Details : Weight : 0.01000kg
Specifications
DNAJC27 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMEANMPK RKEPGRSLRI KVISMGNAEV GKSCIIKRYC EKRFVSKYLA TIGIDYGVTK VHVRDREIKV NIFDMAGHPF FYEVRNEFYK DTQGVILVYD VGQKDSFDAL DAWLAEMKQE LGPHGNMENI IFVVCANKID CTKHRCVDES EGRLWAESKG FLYFETSAQT GEGINEMFQT FYISIVDLCE NGGKRPTTNS SASFTKEQAD AIRRIRNSKD SWDMLGVKPG ASRDEVNKAY RKLAVLLHPD KCVAPGSEDA FKAVVNARTA LLKNIK | |
Greater than 90% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The DNAJC27 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol, 2mM DTT and 2mM EDTA. |