missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DRAXIN Protein

A DNA sequence encoding the Homo sapiens (Human) DRAXIN, was expressed in the hosts and tags indicated.

€ 245.00 - € 3405.00

Specifications

Accession Number NP_940947.3
Gene ID (Entrez) 374946
Name DRAXIN Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16021201
View Documents Promotion Details
enQuireBio™
QP7855-EC-100UG
100 μg
€ 770.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16041201
View Documents Promotion Details
enQuireBio™
QP7855-EC-1MG
1 mg
€ 3405.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16031201
View Documents Promotion Details
enQuireBio™
QP7855-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16051201
View Documents Promotion Details
enQuireBio™
QP7855-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16061201
View Documents Promotion Details
enQuireBio™
QP7855-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16071201
View Documents Promotion Details
enQuireBio™
QP7855-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_940947.3
DRAXIN Protein
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
374946
Research Use Only
DRAXIN
Recombinant Protein
E. coli
GALAPGTPAR NLPENHIDLP GPALWTPQAS HHRRRGPGKK EWGPGLPSQA QDGAVVTATR QASRLPEAEG LLPEQSPAGL LQDKDLLLGL ALPYPEKENR PPGWERTRKR SREHKRRRDR LRLHQGRALV RGPSSLMKKA ELSEAQVLDA AMEESSTSLA PTMFFLTTFE AAPATEESLI LPVTSLRPQQ AQPRSDGEVM PTLDMALFDW TDYEDLKPDG WPSAKKKEKH RGKLSSDGNE TSPAEGEPCD HHQDCLPGTC CDLREHLCTP HNRGLNNKCF DDCMCVEGLR CYAKFHRNRR VTRRKGRCVE PETANGDQGS FINV
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications