missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human DUSP23 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
Description
A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-150 of Human DUSP23 The Recombinant Human DUSP23 Protein is derived from E. coli. The Recombinant Human DUSP23 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications
Specifications
| Concentration | 1mg/mL |
| For Use With (Application) | Functional Study, SDS-PAGE |
| Formulation | Liquid. 20mM Tris-HCl buffer (pH 8.0) containing 2mM DTT, 10% glycerol, 100mM NaCl |
| Gene ID (Entrez) | 54935 |
| Molecular Weight (g/mol) | 18.8 kDa |
| Name | Recombinant Human DUSP23 Protein |
| Purification Method | >90%, by SDS-PAGE |
| Quantity | 10 μg |
| Source | E.coli |
| Storage Requirements | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
| Show More |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction