missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Dystonin Protein

A DNA sequence encoding the Homo sapiens (Human) Dystonin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7837-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16061091

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001138241.1
Dystonin Protein
Research Use Only
DST
Recombinant Protein
E. coli
MAGYLSPAAY LYVEEQEYLQ AYEDVLERYK DERDKVQKKT FTKWINQHLM KVRKHVNDLY EDLRDGHNLI SLLEVLSGDT LPREKGRMRF HRLQNVQIAL DYLKRRQVKL VNIRNDDITD GNPKLTLGLI WTIILHFQIS DIHVTGESED MSAKERLLLW TQQATEGYAG IRCENFTTCW RDGKLFNAII HKYRP
Greater than 90% as determined by SDS-PAGE.
667
50 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers