missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EG-VEGF / prokineticin-1 Protein

Product Code. 15995739
Change view
Click to view available options
Quantity:
1 mg
500 μg
Unit Size:
1mg
500µg
Product Code. Quantity unitSize
15995739 1 mg 1mg
15905749 500 μg 500µg
2 options
This item is not returnable. View return policy

Product Code. 15995739

Brand: enQuireBio™ QP52301mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number NP_115790.1
Gene ID (Entrez) 84432
Name EG-VEGF / prokineticin-1 Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol PROK1
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence AVITGACERD . VQCGAGTCC A. ISLWLRGL RM. CTPLGRE GEE. CHPGSH KVPF. FRKRK HHTCP. CLPN LLCSRF. PDG RYRCSMD. LK NINF
Buffer Sterile filtered (0.22 um) solution containing 0.1% TFA
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.