missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EIF3E Protein

Product Code. 15989029
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15989029 100 μg 100µg
15999029 10 μg 10µg
15939039 50 μg 50µg
15919039 200 μg 200µg
15929039 500 μg 500µg
15909039 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15989029 Supplier enQuireBio™ Supplier No. QP5971ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) EIF3E, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001559.1
Gene ID (Entrez) 3646
Name EIF3E Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol EIF3E
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag GST
Sequence AEYDLTTRIA HFLDRHLVFP LLEFLSVKEI YNEKELLQGK LDLLSDTNMV DFAMDVYKNL YSDDIPHALR EKRTTVVAQL KQLQAETEPI VKMFEDPETT RQMQSTRDGR MLFDYLADKH GFRQEYLDTL YRYAKFQYEC GNYSGAAEYL YFFRVLVPAT DRNALSSLWG KLASEILMQN WDAAMEDLTR LKETIDNNSV SSPLQSLQQR TWLIHWSLFV FFNHPKGRDN IIDLFLYQPQ YLNAIQTMCP HILRYLTTAV ITNKDVRKRR QVLKDLVKVI QQESYTYKDP ITEFVECLYV NFDFDGAQKK LRECESVLVN DFFLVACLED FIENARLFIF ETFCRIHQCI SINMLADKLN MTPEEAERWI VNLIRNARLD AKIDSKLGHV VMGNNAVSPY QQVIEKTKSL SFRSQMLAMN IEKKLNQNSR SEAPNWATQD SGFY
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.