missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human EMMPRIN/CD147 Fc Alexa Fluor™ 488 Protein
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
Description
Measured by flow cytometry for its ability to bind anti-human EMMPRIN/CD147 Monoclonal Antibody conjugated beads.The concentration of Recombinant Human EMMPRIN/CD147 Fc Chimera Alexa Fluor™ 488 (Catalog# AFG972) that produces 50% of the binding response is 10.0-100 ng/mL.
Specifications
Specifications
| Accession Number | NP_001309172.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 682 |
| Molecular Weight (g/mol) | M.W.-observed: 56-69 kDa, under reducing conditions., M.W.-theroretical: 47 kDa (monomer) |
| Quantity | 20 μg |
| Source | Mouse myeloma cell line, NS0-derived human EMMPRIN/CD147 protein Human EMMPRIN/CD147 (Thr25-His205) (N-terminus) Accession # NP_001309172.1 DIEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus) |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | 5F7, basigin, basigin (Ok blood group), BSG, CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction