missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Epididymal secretory glutathione peroxidase Protein

A DNA sequence encoding the Homo sapiens (Human) Epididymal secretory glutathione peroxidase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6116-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16040920

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001500.1
Epididymal secretory glutathione peroxidase Protein
Research Use Only
GPX5
Recombinant Protein
E. coli
MTTQLRVVHL LPLLLACFVQ TSPKQEKMKM DCHKDEKGTI YDYEAIALNK NEYVSFKQYV GKHILFVNVA TYCGLTAQYP GMSVQGEDLY LVSSFLRKGM
Greater than 80% as determined by SDS-PAGE.
2880
200 μg
Not determined.
Full Length of Isoform 2
Human
His-Trx
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers