missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Epididymal secretory glutathione peroxidase Protein
A DNA sequence encoding the Homo sapiens (Human) Epididymal secretory glutathione peroxidase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6116-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001500.1 | |
Epididymal secretory glutathione peroxidase Protein | |
Research Use Only | |
GPX5 | |
Recombinant Protein | |
E. coli | |
MTTQLRVVHL LPLLLACFVQ TSPKQEKMKM DCHKDEKGTI YDYEAIALNK NEYVSFKQYV GKHILFVNVA TYCGLTAQYP GMSVQGEDLY LVSSFLRKGM | |
Greater than 80% as determined by SDS-PAGE. |
2880 | |
50 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-Trx | |
Tris-based buffer, 50% glycerol |