missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Estradiol 17-beta-dehydrogenase 11 Protein

A DNA sequence encoding the Homo sapiens (Human) Estradiol 17-beta-dehydrogenase 11, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7921-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031601

  • € 2385.00 / 1mg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_057329.3
Estradiol 17-beta-dehydrogenase 11 Protein
Research Use Only
HSD17B11
Recombinant Protein
E. coli
ESFVKLFIPK RRKSVTGEIV LITGAGHGIG RLTAYEFAKL KSKLVLWDIN KHGLEETAAK CKGLGAKVHT FVVDCSNRED IYSSAKKVKA EIGDVSILVN NAGVVYTSDL FATQDPQIEK TFEVNVLAHF WTTKAFLPAM TKNNHGHIVT VASAAGHVSV PFLLAYCSSK FAAVGFHKTL TDELAALQIT GVKTTCLCPN FVNTGFIKNP STSLGPTLEP EEVVNRLMHG ILTEQKMIFI PSSIAFLTTL ERILPERFLA VLKQKISVKF DAVIGYKMKA Q
Greater than 90% as determined by SDS-PAGE.
51170
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers