missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EXOSC10 Protein

Product Code. 15918478
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
This item is not returnable. View return policy

Product Code. 15918478

Brand: enQuireBio™ QP118181mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the EXOSC10 was constructed and used to recombinantly synthesize the protein.

Specifications

Name EXOSC10 Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species Baculovirus
Protein Tag His
Sequence MAPPSTREPR VLSATSATKS DGEMVLPGFP DADSFVKFAL GSVVAVTKAS GGLPQFGDEY DFYRSFPGFQ AFCETQGDRL LQCMSRVMQY HGCRSNIKDR SKVTELEDKF DLLVDANDVI LERVGILLDE ASGVNKNQQP VLPAGLQVPK TVVSSWNRKA AEYGKKAKSE TFRLLHAKNI IRPQLKFREK IDNSNTPFLP KIFIKPNAQK PLPQALSKER RERPQDRPED LDVPPALADF IHQQRTQQVE QDMFAHPYQY ELNHFTPADA VLQKPQPQLY RPIEETPCHF ISSLDELVEL NEKLLNCQEF AVDLEHHSYR SFLGLTCLMQ ISTRTEDFII DTLELRSDMY ILNESLTDPA IVKVFHGADS DIEWLQKDFG LYVVNMFDTH QAARLLNLGR HSLDHLLKLY CNVDSNKQYQ LADWRIRPLP EEMLSYARDD THYLLYIYDK MRLEMWERGN GQPVQLQVVW QRSRDICLKK FIKPIFTDES YLELYRKQKK HLNTQQLTAF QLLFAWRDKT ARREDESYGY VLPNHMMLKI AEELPKEPQG IIACCNPVPP LVRQQINEMH LLIQQAREMP LLKSEVAAGV KKSGPLPSAE RLENVLFGPH DCSHAPPDGY PIIPTSGSVP VQKQASLFPD EKEDNLLGTT CLIATAVITL FNEPSAEDSK KGPLTVAQKK AQNIMESFEN PFRMFLPSLG HRAPVSQAAK FDPSTKIYEI SNRWKLAQVQ VQKDSKEAVK KKAAEQTAAR EQAKEACKAA AEQAISVRQQ VVLENAAKKR ERATSDPRTT EQKQEKKRLK ISKKPKDPEP PEKEFTPYDY SQSDFKAFAG NSKSKVSSQF DPNKQTPSGK KCIAAKKIKQ SVGNKSMSFP TGKSDRGFRY NWPQR
Buffer EXOSC10 is supplied at a concentration of 0.8 mg/ml in 20mM HEPES, pH 7.5, 100mM NaCl, 0.01mM EDTA, 0.02% SDS.
Purity or Quality Grade Greater than 70% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.