missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human F Protein

A cDNA sequence encoding the F was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11825-25ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15938498

  • € 181.00 / 25µg

Estimated Shipment: 06-02-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

F Protein
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The Frataxin solution contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT and 10% glycerol.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MLRTDIDATC TPRRASSNQR GLNQIWNVKK QSVYLMNLRK SGTLGHPGSL DETTYERLAE ETLDSLAEFF EDLADKPYTF EDYDVSFGSG VLTVKLGGDL GTYVINKQTP NKQIWLSSPS SGPKRYDWTG KNWVYSHDGV SLHELLAAEL TKALKTKLDL SSLAYSGKDA
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers